Learn More
Medchemexpress LLC 10-undecenal | 112-45-8 | MFCD00007032 | 98.4% | 168.28 g/mol | C11H20O | 10 G
SDP
Catalog No. 5000301330
Encompass_Preferred
10-Undecenal is an unsaturated aliphatic aldehyde (undecylenic aldehyde) with an 11-carbon chain, a terminal aldehyde, and a double bond between the 9th and 10th carbon atoms. It is used as a reagent and building block in organic synthesis and as an analytical standard for method development and characterization.
  • High purity (≈98.4%) for synthetic and analytical applications.
  • Molecular weight 168.28 g/mol; formula C11H20O.
  • Good solubility in DMSO (≥200 mg/mL) for solution-phase work.
  • Terminal aldehyde functionality enables diverse derivatization and coupling reactions.
  • Available in small laboratory pack sizes suitable for R&D use.
Catalog No. 50-003-01330 Supplier Medchemexpress LLC Supplier No. HYW01882910G
10-Undecenal is an unsaturated aliphatic aldehyde (undecylenic aldehyde) with an 11-carbon chain, a terminal aldehyde, and a double bond between the 9th and 10th carbon atoms. It is used as a reagent and building block in organic synthesis and as an analytical standard for method development and characterization.
  • High purity (≈98.4%) for synthetic and analytical applications.
  • Molecular weight 168.28 g/mol; formula C11H20O.
  • Good solubility in DMSO (≥200 mg/mL) for solution-phase work.
  • Terminal aldehyde functionality enables diverse derivatization and coupling reactions.
  • Available in small laboratory pack sizes suitable for R&D use.