Learn More
Medchemexpress LLC 10-undecenal | 112-45-8 | MFCD00007032 | 98.4% | 168.28 g/mol | C11H20O | 25 G
SDP
Catalog No. 5000302830
Encompass_Preferred
10-Undecenal (undecylenic aldehyde) is an 11-carbon aliphatic aldehyde used as a reagent and intermediate in organic synthesis, fragrance and flavor chemistry, and analytical applications. It is supplied as a colorless to light-yellow liquid with a typical purity near 98% and a molecular weight of 168.28 g/mol.
  • High purity suitable for synthetic and analytical work.
  • Liquid form simplifies handling and metering.
  • Compatible with common organic solvents; store under inert atmosphere.
  • Useful as a building block for aldehyde-containing intermediates.
  • Batch-specific certificate of analysis available for quality verification.
Catalog No. 50-003-02830 Supplier Medchemexpress LLC Supplier No. 25GHYW018829
10-Undecenal (undecylenic aldehyde) is an 11-carbon aliphatic aldehyde used as a reagent and intermediate in organic synthesis, fragrance and flavor chemistry, and analytical applications. It is supplied as a colorless to light-yellow liquid with a typical purity near 98% and a molecular weight of 168.28 g/mol.
  • High purity suitable for synthetic and analytical work.
  • Liquid form simplifies handling and metering.
  • Compatible with common organic solvents; store under inert atmosphere.
  • Useful as a building block for aldehyde-containing intermediates.
  • Batch-specific certificate of analysis available for quality verification.