Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 10-undecenoic acid | 112-38-9 | MFCD00004442 | 100.0% | 184.28 | C11H20O2 | 10 MG
SDP

Catalog No. 5000297380
Encompass_Preferred

Analytical reference standard of 10-undecenoic acid (undecylenic acid) intended for research and analytical use. High-purity material is suitable for method development, calibration, and compound identification in chromatographic and spectrometric assays; not for human or clinical use.

  • High purity reported as 99.99%.
  • CAS number 112-38-9 and formula C11H20O2.
  • Molecular weight 184.28 g/mol.
  • Provided as a small reference pack (10 mg) for analytical workflows.
  • Functions as an antifungal agent and is reported to inhibit Aβ oligomerization and scavenge reactive oxygen species.
  • Intended for research use only; not for patient use.

Catalog No. 50-002-97380 Supplier Medchemexpress LLC Supplier No. HYB0914R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Analytical reference standard of 10-undecenoic acid (undecylenic acid) intended for research and analytical use. High-purity material is suitable for method development, calibration, and compound identification in chromatographic and spectrometric assays; not for human or clinical use.

  • High purity reported as 99.99%.
  • CAS number 112-38-9 and formula C11H20O2.
  • Molecular weight 184.28 g/mol.
  • Provided as a small reference pack (10 mg) for analytical workflows.
  • Functions as an antifungal agent and is reported to inhibit Aβ oligomerization and scavenge reactive oxygen species.
  • Intended for research use only; not for patient use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.