Learn More
Medchemexpress LLC 10-Undecenoic acid | 112-38-9 | MFCD00004442 | 97.3% | 100 MG
SDP
Catalog No. 5000407692
Encompass_Preferred
10-Undecenoic acid (Undecylenic acid) is an antifungal agent. It inhibits Aβ oligomerization, scavenges ROS, and inhibits μ-calpain activity. It has neuroprotective and anticancer effects on various tumors. Additionally, it inhibits C. albicans biofilm formation and MRSA infection, and suppresses quorum sensing signals of Bacillus subtilis and Pseudomonas aeruginosa.
  • Antifungal agent
  • Inhibits Aβ oligomerization
  • Scavenges reactive oxygen species (ROS)
  • Inhibits μ-calpain activity
  • Possesses neuroprotective effects
  • Exhibits anticancer effects on various tumors
  • Inhibits C. albicans biofilm formation and MRSA infection
  • Inhibits quorum sensing signals of Bacillus subtilis and Pseudomonas aeruginosa
Catalog No. 50-004-07692 Supplier Medchemexpress LLC Supplier No. HYB0914R100MG
10-Undecenoic acid (Undecylenic acid) is an antifungal agent. It inhibits Aβ oligomerization, scavenges ROS, and inhibits μ-calpain activity. It has neuroprotective and anticancer effects on various tumors. Additionally, it inhibits C. albicans biofilm formation and MRSA infection, and suppresses quorum sensing signals of Bacillus subtilis and Pseudomonas aeruginosa.
  • Antifungal agent
  • Inhibits Aβ oligomerization
  • Scavenges reactive oxygen species (ROS)
  • Inhibits μ-calpain activity
  • Possesses neuroprotective effects
  • Exhibits anticancer effects on various tumors
  • Inhibits C. albicans biofilm formation and MRSA infection
  • Inhibits quorum sensing signals of Bacillus subtilis and Pseudomonas aeruginosa