Learn More
Medchemexpress LLC 10-Undecenoic acid | 112-38-9 | MFCD00004442 | 99.99% | 100 G
SDP
Catalog No. 5000409346
Encompass_Preferred
10-Undecenoic acid, also known as Undecylenic acid, is a potent antifungal agent. It demonstrates neuroprotective effects by inhibiting Aβ oligomerization, scavenging reactive oxygen species, and inhibiting μ-calpain activity. Additionally, it exhibits anticancer effects across various tumor types and plays a role in inhibiting biofilm formation and quorum sensing signals.
  • Acts as an antifungal agent.
  • Inhibits Aβ oligomerization and scavenges reactive oxygen species.
  • Demonstrates neuroprotective effects.
  • Exhibits anticancer effects on various tumors.
  • Inhibits C. albicans biofilm formation and MRSA infection.
  • Suppresses quorum sensing signals of Bacillus subtilis and Pseudomonas aeruginosa.
Catalog No. 50-004-09346 Supplier Medchemexpress LLC Supplier No. HYB0914100G
10-Undecenoic acid, also known as Undecylenic acid, is a potent antifungal agent. It demonstrates neuroprotective effects by inhibiting Aβ oligomerization, scavenging reactive oxygen species, and inhibiting μ-calpain activity. Additionally, it exhibits anticancer effects across various tumor types and plays a role in inhibiting biofilm formation and quorum sensing signals.
  • Acts as an antifungal agent.
  • Inhibits Aβ oligomerization and scavenges reactive oxygen species.
  • Demonstrates neuroprotective effects.
  • Exhibits anticancer effects on various tumors.
  • Inhibits C. albicans biofilm formation and MRSA infection.
  • Suppresses quorum sensing signals of Bacillus subtilis and Pseudomonas aeruginosa.