Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 10-Undecenoic acid

Catalog No. 124672500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-467-2500ML 500 mL
12-467-2100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-467-2500ML Supplier Sigma Aldrich Supplier No. 124672500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

10-Undecenoic acid

Specifications

CAS 112-38-9
Melting Point 23°C to 25°C (lit.)
Density 0.912 g/mL (at 25°C (literature))
Boiling Point 137°C (2 mmHg)
Molecular Formula C11H20O2
Linear Formula CH2=CH(CH2)8 COOH
Refractive Index n20/D 1.449 (literature)
MDL Number MFCD00004442
Quantity 500 mL
Synonym Undecylenic acid
Molecular Weight (g/mol) 184.28
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.