Learn More
Medchemexpress LLC 10-Undecenoic acid (stabilized with TBC) | 112-38-9 | 100.0% | 1 G
SDP
Catalog No. 5000408472
Encompass_Preferred
10-Undecenoic acid (Undecylenic acid) is an antifungal agent. It inhibits Aβ oligomerization, scavenges reactive oxygen species (ROS), and inhibits μ-calpain activity. This compound also exhibits neuroprotective effects and anticancer effects on various tumors. Additionally, 10-Undecenoic acid inhibits *C. albicans* biofilm formation and MRSA infection. It also inhibits quorum sensing signals of *Bacillus subtilis* and *Pseudomonas aeruginosa*.
  • Acts as an antifungal agent.
  • Inhibits Aβ oligomerization.
  • Scavenges reactive oxygen species (ROS).
  • Inhibits μ-calpain activity.
  • Exhibits neuroprotective effects.
  • Exhibits anticancer effects on various tumors.
  • Inhibits *C. albicans* biofilm formation and MRSA infection.
  • Inhibits quorum sensing signals of *Bacillus subtilis* and *Pseudomonas aeruginosa*.
Catalog No. 50-004-08472 Supplier Medchemexpress LLC Supplier No. HYB09141G
10-Undecenoic acid (Undecylenic acid) is an antifungal agent. It inhibits Aβ oligomerization, scavenges reactive oxygen species (ROS), and inhibits μ-calpain activity. This compound also exhibits neuroprotective effects and anticancer effects on various tumors. Additionally, 10-Undecenoic acid inhibits *C. albicans* biofilm formation and MRSA infection. It also inhibits quorum sensing signals of *Bacillus subtilis* and *Pseudomonas aeruginosa*.
  • Acts as an antifungal agent.
  • Inhibits Aβ oligomerization.
  • Scavenges reactive oxygen species (ROS).
  • Inhibits μ-calpain activity.
  • Exhibits neuroprotective effects.
  • Exhibits anticancer effects on various tumors.
  • Inhibits *C. albicans* biofilm formation and MRSA infection.
  • Inhibits quorum sensing signals of *Bacillus subtilis* and *Pseudomonas aeruginosa*.