Learn More
Medchemexpress LLC 10-Undecenoic acid, stabilized with TBC | 112-38-9 | 98% | 1 G
SDP
Catalog No. 5000409222
Encompass_Preferred
10-Undecenoic acid, also known as Undecylenic acid, is an antifungal agent that also exhibits properties such as inhibiting Aβ oligomerization, scavenging reactive oxygen species (ROS), and inhibiting μ-calpain activity. This chemical is stabilized with TBC and is intended for research use only.
  • Antifungal agent
  • Inhibits Aβ oligomerization
  • Scavenges reactive oxygen species (ROS)
  • Inhibits μ-calpain activity
  • Stabilized with TBC
  • For research use only
  • Storage conditions:
    • Pure form: -20°C for 3 years; 4°C for 2 years
    • In solvent: -80°C for 6 months; -20°C for 1 month
    • Shipping at room temperature if less than 2 weeks
Catalog No. 50-004-09222 Supplier Medchemexpress LLC Supplier No. HYB0914A1G
10-Undecenoic acid, also known as Undecylenic acid, is an antifungal agent that also exhibits properties such as inhibiting Aβ oligomerization, scavenging reactive oxygen species (ROS), and inhibiting μ-calpain activity. This chemical is stabilized with TBC and is intended for research use only.
  • Antifungal agent
  • Inhibits Aβ oligomerization
  • Scavenges reactive oxygen species (ROS)
  • Inhibits μ-calpain activity
  • Stabilized with TBC
  • For research use only
  • Storage conditions:
    • Pure form: -20°C for 3 years; 4°C for 2 years
    • In solvent: -80°C for 6 months; -20°C for 1 month
    • Shipping at room temperature if less than 2 weeks