Learn More
Medchemexpress LLC 10-Undecenoic acid (stabilized with TBC) | 112-38-9 | 98% | 25 MG
SDP
Catalog No. 5000450727
Encompass_Preferred
10-Undecenoic acid (Undecylenic acid) is an antifungal agent that inhibits Aβ oligomerization, scavenges Reactive Oxygen Species (ROS), and inhibits μ-calpain activity. It exhibits neuroprotective and anticancer effects on various tumors. This compound also inhibits *C. albicans* biofilm formation, MRSA infection, and quorum sensing signals of *Bacillus subtilis* and *Pseudomonas aeruginosa*.
  • Antifungal agent
  • Inhibits Aβ oligomerization
  • Scavenges reactive oxygen species (ROS)
  • Inhibits μ-calpain activity
  • Neuroprotective effects
  • Anticancer effects on various tumors
  • Inhibits *C. albicans* biofilm formation
  • Inhibits MRSA infection
  • Inhibits quorum sensing signals of *Bacillus subtilis* and *Pseudomonas aeruginosa*
Catalog No. 50-004-50727 Supplier Medchemexpress LLC Supplier No. 25MGHYB0914R
10-Undecenoic acid (Undecylenic acid) is an antifungal agent that inhibits Aβ oligomerization, scavenges Reactive Oxygen Species (ROS), and inhibits μ-calpain activity. It exhibits neuroprotective and anticancer effects on various tumors. This compound also inhibits *C. albicans* biofilm formation, MRSA infection, and quorum sensing signals of *Bacillus subtilis* and *Pseudomonas aeruginosa*.
  • Antifungal agent
  • Inhibits Aβ oligomerization
  • Scavenges reactive oxygen species (ROS)
  • Inhibits μ-calpain activity
  • Neuroprotective effects
  • Anticancer effects on various tumors
  • Inhibits *C. albicans* biofilm formation
  • Inhibits MRSA infection
  • Inhibits quorum sensing signals of *Bacillus subtilis* and *Pseudomonas aeruginosa*