Learn More
Medchemexpress LLC 10-Undecenoic acid (Standard) | 112-38-9 | 97.3% | 50 MG
SDP
Catalog No. 5000409290
Encompass_Preferred
10-Undecenoic acid (Standard) is an analytical standard and an antifungal agent. It is intended for research and analytical applications, serving as a reference for assays. This compound is utilized in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS.
  • Inhibits Aβ oligomerization
  • Scavenges ROS
  • Inhibits μ-calpain activity, exhibiting neuroprotective effects
  • Exhibits anticancer effects on various tumors
  • Inhibits *C. albicans* biofilm formation and MRSA infection
  • Suppresses quorum sensing signals of *Bacillus subtilis* and *Pseudomonas aeruginosa*
Catalog No. 50-004-09290 Supplier Medchemexpress LLC Supplier No. HYB0914R50MG
10-Undecenoic acid (Standard) is an analytical standard and an antifungal agent. It is intended for research and analytical applications, serving as a reference for assays. This compound is utilized in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS.
  • Inhibits Aβ oligomerization
  • Scavenges ROS
  • Inhibits μ-calpain activity, exhibiting neuroprotective effects
  • Exhibits anticancer effects on various tumors
  • Inhibits *C. albicans* biofilm formation and MRSA infection
  • Suppresses quorum sensing signals of *Bacillus subtilis* and *Pseudomonas aeruginosa*