Learn More
Medchemexpress LLC 10-Undecenoic acid (Undecylenic acid) | 112-38-9 | 100.0% | 50 G
SDP
Catalog No. 5000407671
Encompass_Preferred
10-Undecenoic acid (Undecylenic acid) is an antifungal agent with neuroprotective and anticancer effects. It works by inhibiting Aβ oligomerization, scavenging Reactive Oxygen Species (ROS), and inhibiting μ-calpain activity. This compound has shown efficacy against *C. albicans* biofilm formation and MRSA infection, and can also inhibit quorum sensing signals in *Bacillus subtilis* and *Pseudomonas aeruginosa*.
  • Induces apoptosis in various tumor cells
  • Stimulates osteoblast differentiation
  • Offers neuroprotection against cell death induced by glutamate, H2O2, and Aβ1-42
  • Inhibits *C. albicans* biofilm formation and MRSA infection
  • Scavenges reactive oxygen species
Catalog No. 50-004-07671 Supplier Medchemexpress LLC Supplier No. HYB091450G
10-Undecenoic acid (Undecylenic acid) is an antifungal agent with neuroprotective and anticancer effects. It works by inhibiting Aβ oligomerization, scavenging Reactive Oxygen Species (ROS), and inhibiting μ-calpain activity. This compound has shown efficacy against *C. albicans* biofilm formation and MRSA infection, and can also inhibit quorum sensing signals in *Bacillus subtilis* and *Pseudomonas aeruginosa*.
  • Induces apoptosis in various tumor cells
  • Stimulates osteoblast differentiation
  • Offers neuroprotection against cell death induced by glutamate, H2O2, and Aβ1-42
  • Inhibits *C. albicans* biofilm formation and MRSA infection
  • Scavenges reactive oxygen species