Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 10-Undecenoyl chloride

Catalog No. 16166725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-166-725G 25 g
16-166-7100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-166-725G Supplier Sigma Aldrich Supplier No. 16166725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 10-undecenoyl chloride was used as acylating reagent in the synthesis of cellulose -d-carboxyalkanoates and poly(ethylene glycol)-lipid amphiphiles
  • It was used in synthesis and modification of hyperbranched poly(glycidol)

Specifications

Boiling Point 120°C to 122°C (10 mmHg)
CAS 38460-95-6
Quantity 25 g
Molecular Formula C11H19ClO
Linear Formula CH2=CH(CH2)8 COCl
MDL Number MFCD00000772
Molecular Weight (g/mol) 202.72
Percent Purity 97%
Refractive Index n20/D 1.454 (literature)
Density 0.944 g/mL (at 25°C (literature))
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.