Learn More
Teknova 10% (v/v) Antifoam 204. 440mL in 1L Bottle, Sterile.
SDP
Catalog No. 502388173
Encompass
Antifoam Solution is used to reduce foaming during fermentation and manipulation of cultured cells. This helps minimize damage to cells, resulting in increased protein/antibody production.
Catalog No. 50-238-8173 Supplier Teknova Supplier No. A6427
Antifoam Solution is used to reduce foaming during fermentation and manipulation of cultured cells. This helps minimize damage to cells, resulting in increased protein/antibody production.