Learn More
Rockland Immunochemicals SHEEP RED BLOOD CELLS (RBC) 10% Washed Pooled Cells - R405-0050
SDP
Catalog No. NC9914520
Encompass_Preferred
Sheep whole blood is washed to remove the platelet rich plasma, buffy coat layer, and leukocytes (WBC). Red blood cells are supplied as a 10 percent suspension in phosphate buffered saline (PBS). Sheep red blood cells are useful for the titration of complement, adsorption procedures, testing for agglutinins/HA assays, and for the preparation of stroma as particulate reagents. Sheep red blood cells are perishable and are collected and processed upon receipt of your order.
Catalog No. NC9914520 Supplier Rockland Immunochemicals Supplier No. R4050050
Sheep whole blood is washed to remove the platelet rich plasma, buffy coat layer, and leukocytes (WBC). Red blood cells are supplied as a 10 percent suspension in phosphate buffered saline (PBS). Sheep red blood cells are useful for the titration of complement, adsorption procedures, testing for agglutinins/HA assays, and for the preparation of stroma as particulate reagents. Sheep red blood cells are perishable and are collected and processed upon receipt of your order.