Learn More
Euthanex 10 X 16 LID WITH FLOW RESTRICT
Catalog No. NC2319287
Encompass
Small Euthanex Lid, to be used in conjunction with cages sized to fit within these dimensions for a tight seal. Euthanex lids allow animals to be gently euthanized through the precise control of gas introduction. The animals remain in their home cages, thereby minimizing added stress. The lid system is highly economical and efficient and still widely used today. Our lids are constructed of heavy-duty, stainless steel. They include an easy, quick disconnect lid stem for gas input as well as a foam gasket for a secure fit. The foam gasket seal allows the lids to be used with smaller cage sizes.
Catalog No. NC2319287 Supplier Euthanex Supplier No. E20028R
Small Euthanex Lid, to be used in conjunction with cages sized to fit within these dimensions for a tight seal. Euthanex lids allow animals to be gently euthanized through the precise control of gas introduction. The animals remain in their home cages, thereby minimizing added stress. The lid system is highly economical and efficient and still widely used today. Our lids are constructed of heavy-duty, stainless steel. They include an easy, quick disconnect lid stem for gas input as well as a foam gasket for a secure fit. The foam gasket seal allows the lids to be used with smaller cage sizes.