Learn More
Ace Glass, Inc. 10 x 20mm porosity B stick GLASSBLOWER
SDP
Catalog No. 50978177
Encompass_Preferred
10mm O.D. x 20mm long porous fritted (70-100 micron) glass filter stick, used to fabricate gas disperstion tubes. Hollow at one end. Pack of 6.
Catalog No. 50-978-177 Supplier Ace Glass, Inc. Supplier No. 1533705
10mm O.D. x 20mm long porous fritted (70-100 micron) glass filter stick, used to fabricate gas disperstion tubes. Hollow at one end. Pack of 6.