Learn More
Genscript Corporation 10 x iTaq/i Buffer
Catalog No. 5000058347
Encompass
The 10 x i Taq /i Buffer contains 500 mM KCl 100 mM Tris HCl (pH 9 0 at 25 C) 15 mM MgCl sub 2 /sub and 1% Triton X-100 This buffer is optimized for use with 200 M dNTPs
Catalog No. 50-000-58347 Supplier Genscript Corporation Supplier No. B0005
The 10 x i Taq /i Buffer contains 500 mM KCl 100 mM Tris HCl (pH 9 0 at 25 C) 15 mM MgCl sub 2 /sub and 1% Triton X-100 This buffer is optimized for use with 200 M dNTPs