Learn More
Teknova 10X TBE, DNA Sequencing Grade Solution. 4L, Non-Sterile.
SDP
Supplier:  Teknova T0210
Encompass
890mM Tris, 890mM Boric Acid, 20mM Ethylenediaminetetraacetic acid (EDTA)
Catalog No. 50-238-8061