Learn More
Cayman Chemical 10(Z)-Pentadecenoic Acid, Purity: ≥98%, CAS: 84743-29-3, 25 mg
SDP
Catalog No. 502964345
Encompass_Preferred
A 15-carbon monounsaturated fatty acid; inhibits IFN-γ-induced production of kynurenine in Thp-1 cells by 16%
Catalog No. 50-296-4345 Supplier Cayman Chemical Supplier No. 2236225 MG
A 15-carbon monounsaturated fatty acid; inhibits IFN-γ-induced production of kynurenine in Thp-1 cells by 16%