Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 10(Z)-pentadecenoic acid | 84743-29-3 | 99.9% | 1 MG
SDP

Catalog No. 5000233992
Encompass_Preferred

10(Z)-pentadecenoic acid is a monounsaturated long-chain fatty acid (cis-10 isomer) used as a biochemical reagent and analytical reference in lipid research.

  • High purity (99.86%).
  • Molecular formula C15H28O2 and molecular weight 240.38 g/mol.
  • Soluble in DMSO at 100 mg/mL; may require sonication.
  • Store protected from light; in solution: -80°C (6 months), -20°C (1 month).
  • Available in small mass quantities suitable for analytical use.

Catalog No. 50-002-33992 Supplier Medchemexpress LLC Supplier No. HY120996B1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

10(Z)-pentadecenoic acid is a monounsaturated long-chain fatty acid (cis-10 isomer) used as a biochemical reagent and analytical reference in lipid research.

  • High purity (99.86%).
  • Molecular formula C15H28O2 and molecular weight 240.38 g/mol.
  • Soluble in DMSO at 100 mg/mL; may require sonication.
  • Store protected from light; in solution: -80°C (6 months), -20°C (1 month).
  • Available in small mass quantities suitable for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.