Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 10(Z)-pentadecenoic acid | 84743-29-3 | 99.9% | 5 MG
SDP

Catalog No. 5000233993
Encompass_Preferred

10(Z)-Pentadecenoic acid is a long-chain monounsaturated fatty acid used as a research reagent in lipid and biochemical studies.

  • Long-chain monounsaturated fatty acid (15 carbons).
  • Molecular weight 240.38 g/mol.
  • Chemical formula C15H28O2.
  • High purity (≈99.9%).
  • Supplied in small milligram quantities for research use.
  • Store refrigerated and protect from light; in solvent, -80°C for up to 6 months, -20°C for up to 1 month.

Catalog No. 50-002-33993 Supplier Medchemexpress LLC Supplier No. HY120996B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

10(Z)-Pentadecenoic acid is a long-chain monounsaturated fatty acid used as a research reagent in lipid and biochemical studies.

  • Long-chain monounsaturated fatty acid (15 carbons).
  • Molecular weight 240.38 g/mol.
  • Chemical formula C15H28O2.
  • High purity (≈99.9%).
  • Supplied in small milligram quantities for research use.
  • Store refrigerated and protect from light; in solvent, -80°C for up to 6 months, -20°C for up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.