Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab 10% Zinc Formalin Fixative
SDP

Catalog No. 21060018
Change view
Click to view available options
Form:
Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
21-060-018 Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-060-018 Supplier StatLab Supplier No. FXZFOGAL
Harmful if swallowed, in contact with skin, or if inhaled

A primary fixative used in histology to preserve tissues for further analysis.

  • Solution is an environmentally safer alternative to traditional fixatives like Mercuric Chloride
  • This fixative works by altering protein charges and inducing cross-linking, which helps maintain the structural integrity of tissues
  • Compatible with various staining techniques and can be used in all tissue processors

Specifications

For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Fixative
Quantity 1 gal.
Solution Type Fixative
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.