Learn More
Cayman Chemical 10(Z)-Nonadecenoic Acid methyl ester, Purity: ≥98%, CAS: 19788-74-0, 250 mg
SDP
Catalog No. 502935335
Encompass_Preferred
A fatty acid methyl ester; has been used as a standard for the quantification of cis-nonadecenoic acid in chromatography applications
Catalog No. 50-293-5335 Supplier Cayman Chemical Supplier No. 26733250 MG
A fatty acid methyl ester; has been used as a standard for the quantification of cis-nonadecenoic acid in chromatography applications