Learn More
Cayman Chemical 10(Z)-Nonadecenoic Acid methyl ester, Purity: ≥98%, CAS: 19788-74-0, 100 mg
SDP
Catalog No. 502934379
Encompass_Preferred
A fatty acid methyl ester; has been used as a standard for the quantification of cis-nonadecenoic acid in chromatography applications
Catalog No. 50-293-4379 Supplier Cayman Chemical Supplier No. 26733100 MG
A fatty acid methyl ester; has been used as a standard for the quantification of cis-nonadecenoic acid in chromatography applications