Learn More
Cayman Chemical 10(Z)-Pentadecenoic Acid methyl ester, Purity: ≥99%, CAS: 90176-52-6, 50 mg
SDP
Catalog No. 502934395
Encompass_Preferred
A methyl ester form of 10(Z)-pentadecenoic acid; has been used as a standard for the quantification of pentadecenoic acid in vernix caseosa
Catalog No. 50-293-4395 Supplier Cayman Chemical Supplier No. 2672950 MG
A methyl ester form of 10(Z)-pentadecenoic acid; has been used as a standard for the quantification of pentadecenoic acid in vernix caseosa