Learn More
Hach Company 100-130 mg/L Chlorine Standard, 20mL
Catalog No. NC1675735 Shop All Hach Company Products
Encompass
This chlorine standard can be diluted down to verify the accuracy of the Cl17 chlorine analyser or create a separate calibration curve. Under normal circumstances, a separate calibration curve is not required for the Cl17. Package of one 20 mL ampule.
Catalog No. NC1675735 Supplier Hach Company Supplier No. 2835801
This chlorine standard can be diluted down to verify the accuracy of the Cl17 chlorine analyser or create a separate calibration curve. Under normal circumstances, a separate calibration curve is not required for the Cl17. Package of one 20 mL ampule.