Learn More
Chemglass Life Sciences 100-440 G.P.I. Glass Thread, 110mm OD Tubing, 175mm OAL
SDP
Catalog No. CG19016
Encompass_Preferred
100-440 G.P.I. Glass Thread, 110mm OD Tubing, 175mm OAL. Thread for replacement on apparatus using G.P.I. (Glass Packaging Institute) thread.PLEASE NOTE: GPI THREADS ARE SOLD IN PACKAGES ONLY.
Catalog No. CG19016 Supplier Chemglass Life Sciences Supplier No. CG19016
100-440 G.P.I. Glass Thread, 110mm OD Tubing, 175mm OAL. Thread for replacement on apparatus using G.P.I. (Glass Packaging Institute) thread.PLEASE NOTE: GPI THREADS ARE SOLD IN PACKAGES ONLY.