Learn More
Chemglass Life Sciences Solid Cap, 100-440 G.P.I. Thread, PTFE Liner
SDP
Catalog No. CG19114
Encompass_Preferred
Solid Cap, 100-440 G.P.I. Thread, PTFE Liner. Cap for use with CG-190 G.P.I. screw threads. Part numbers CG-191-01 through CG-191-12 are made of black plastic with a choice of white rubber or PTFE liner. Part number CG-191-14 is made of blue polypropylene with a detachable PTFE liner. NOTE: CAPS SOLD IN PACKAGES ONLY.
Catalog No. CG19114 Supplier Chemglass Life Sciences Supplier No. CG19114
Solid Cap, 100-440 G.P.I. Thread, PTFE Liner. Cap for use with CG-190 G.P.I. screw threads. Part numbers CG-191-01 through CG-191-12 are made of black plastic with a choice of white rubber or PTFE liner. Part number CG-191-14 is made of blue polypropylene with a detachable PTFE liner. NOTE: CAPS SOLD IN PACKAGES ONLY.