Learn More
Thermco Products Inc NIST compliant, factory certified glass Spirit thermometer, -100 deg./50 deg.C, 1.0C div, TOTAL imm, certified at -78 deg.C, -20 deg.C, 0 deg.C, 4 deg.C, 25 deg.C, 37 deg.C, 305mm
Catalog No. NC0522020
Encompass_Preferred
Thermco Certified Thermometers are used in temperature verification and calibration procedures. These instruments can be used to calibrate Liquid-In- Glass, RTD, Thermistors, Thermocouples, and Bi-Metal Thermometers. The certified thermometers are carefully inspected to insure they conform to exact specifications and are certified against NlST standards. Each thermometer is furnished with a document proving traceability and a felt-lined leatherette storage case.
Catalog No. NC0522020 Supplier Thermco Products Inc Supplier No. ACC603FC
Thermco Certified Thermometers are used in temperature verification and calibration procedures. These instruments can be used to calibrate Liquid-In- Glass, RTD, Thermistors, Thermocouples, and Bi-Metal Thermometers. The certified thermometers are carefully inspected to insure they conform to exact specifications and are certified against NlST standards. Each thermometer is furnished with a document proving traceability and a felt-lined leatherette storage case.