Learn More
Gilson Company Inc 100 ALPINE W/SEAL
Catalog No. NC9203638
Encompass_Preferred
Micron Air Jet Sieve, Size: no.100, (150micrometer), Standard 200mm dia., Stainless Steel Frame and Mesh
Catalog No. NC9203638 Supplier Gilson Company Inc Supplier No. AJA224
Micron Air Jet Sieve, Size: no.100, (150micrometer), Standard 200mm dia., Stainless Steel Frame and Mesh