Learn More
Teknova 100% Antifoam 204. 20mL in 60mL, Sterile. 25 pack.
SDP
Supplier:  Teknova A642625PK
Encompass
Antifoam Solution is used to reduce foaming during fermentation and manipulation of cultured cells. This helps minimize damage to cells, resulting in increased protein/antibody production.
Catalog No. 50-274-5869