Learn More
Teknova 100% Antifoam 204. 500mL, Sterile 8 pack.
SDP
Catalog No. 502745988
Encompass
Antifoam Solution is used to reduce foaming during fermentation and manipulation of cultured cells. This helps minimize damage to cells, resulting in increased protein/antibody production.
Catalog No. 50-274-5988 Supplier Teknova Supplier No. 2A64108PK
Antifoam Solution is used to reduce foaming during fermentation and manipulation of cultured cells. This helps minimize damage to cells, resulting in increased protein/antibody production.