Learn More
Medical Electronic Systems LLC 100/BOTTLE 0700 Q CHK TEST STR
SDP
Catalog No. NC0257987
Encompass
QwikCheck Test Strips is a kit used to determine pH and leukocytes (WBCs) in semen. Test results are determined by comparing the color of the test patches to the color chart provided by the label on the bottle.
Catalog No. NC0257987 Supplier Medical Electronic Systems LLC Supplier No. 0700
QwikCheck Test Strips is a kit used to determine pH and leukocytes (WBCs) in semen. Test results are determined by comparing the color of the test patches to the color chart provided by the label on the bottle.