Learn More
Ubpbio 100 bp DNA Ladder, 500UL
Catalog No. 501965068
Encompass_Preferred
A unique combination of PCR products and a number of proprietary plasmids digested with appropriate restriction enzymes to yield 12 fragments, suitable for use as molecular weight standards for agarose gel electrophoresis. The DNA includes fragments ranging from 100-3,000 base pairs. The 500 and 1,500 base pair bands have increased intensity to serve as reference points. The approximate amount of DNA in each band is provided (0.54 μg per load).
Catalog No. 50-196-5068 Supplier Ubpbio Supplier No. L1010
A unique combination of PCR products and a number of proprietary plasmids digested with appropriate restriction enzymes to yield 12 fragments, suitable for use as molecular weight standards for agarose gel electrophoresis. The DNA includes fragments ranging from 100-3,000 base pairs. The 500 and 1,500 base pair bands have increased intensity to serve as reference points. The approximate amount of DNA in each band is provided (0.54 μg per load).