Learn More
Medchemexpress LLC 100 bp DNA marker | 250 UL
SDP
Supplier:  Medchemexpress LLC 250ULHYK0802
Encompass_Preferred
The 100 bp DNA Marker is provided in a solution of 1x DNA Loading Buffer and can be directly used for nucleic acid electrophoresis analysis. It contains 10 double-stranded DNA fragments ranging from 100 bp to 1000 bp. A 5 μL aliquot of this product contains approximately 120 ng for the 500 bp band and about 50 ng for the other bands, which is useful for quantitative analysis.
  • Provided in 1x DNA loading buffer
  • Directly usable for nucleic acid electrophoresis analysis
  • Contains 10 double-stranded DNA fragments
  • Fragments range from 100 bp to 1000 bp
  • 5 μL of the product contains approximately 120 ng for the 500 bp band and about 50 ng for the other bands
Catalog No. 50-004-30474