Learn More
Medchemexpress LLC 100 bp DNA Marker | 500 UL
SDP
Catalog No. 5000430191
Encompass_Preferred
The 100 bp DNA Marker is provided in a solution of 1x DNA Loading Buffer, designed for direct use in nucleic acid electrophoresis analysis. This marker contains 10 double-stranded DNA fragments, with sizes ranging from 100 bp to 1000 bp. A 5 μL aliquot typically contains approximately 120 ng for the 500 bp band and about 50 ng for the other bands.
  • Supplied in 1x DNA Loading Buffer for direct use
  • Contains 10 double-stranded DNA fragments
  • Fragment sizes range from 100 bp to 1000 bp
  • 5 μL contains approximately 120 ng for 500 bp band
  • 5 μL contains approximately 50 ng for other bands
Catalog No. 50-004-30191 Supplier Medchemexpress LLC Supplier No. HYK0802500UL
The 100 bp DNA Marker is provided in a solution of 1x DNA Loading Buffer, designed for direct use in nucleic acid electrophoresis analysis. This marker contains 10 double-stranded DNA fragments, with sizes ranging from 100 bp to 1000 bp. A 5 μL aliquot typically contains approximately 120 ng for the 500 bp band and about 50 ng for the other bands.
  • Supplied in 1x DNA Loading Buffer for direct use
  • Contains 10 double-stranded DNA fragments
  • Fragment sizes range from 100 bp to 1000 bp
  • 5 μL contains approximately 120 ng for 500 bp band
  • 5 μL contains approximately 50 ng for other bands