Learn More
Gold Biotechnology Inc 100 bp PLUS™ DNA Ladder 500 µL
SDP
Catalog No. 501532758
Encompass_Preferred
A unique combination of PCR products and a number of proprietary plasmids digested with appropriate restriction enzymes to yield 12 fragments, suitable for use as molecular weight standards for agarose gel electrophoresis. The DNA includes fragments ranging from 100-3,000 base pairs. The 500 and 1,500 base pair bands have increased intensity to serve as reference points. The approximate mass of DNA in each band is provided (0.5 μg a load) for approximating the mass of DNA in comparably intense samples of similar size.The ladder is ready-to-use, and is premixed with loading buffer dye for direct loading on gel.
Catalog No. 50-153-2758 Supplier Gold Biotechnology Inc Supplier No. D003500
A unique combination of PCR products and a number of proprietary plasmids digested with appropriate restriction enzymes to yield 12 fragments, suitable for use as molecular weight standards for agarose gel electrophoresis. The DNA includes fragments ranging from 100-3,000 base pairs. The 500 and 1,500 base pair bands have increased intensity to serve as reference points. The approximate mass of DNA in each band is provided (0.5 μg a load) for approximating the mass of DNA in comparably intense samples of similar size.The ladder is ready-to-use, and is premixed with loading buffer dye for direct loading on gel.