Learn More
LAMDA BIOTECH INC 100 bp Quantitative DNA Ladder
Catalog No. 502045809
Encompass_Preferred
100bp Quanti DNA Ladder consists of 9 fragments ranging from 100-1500 base pairs (bp). This molecular weight Marker is specially designed for identifying small DNA fragments and allowing for quantitative estimation of the DNA fragments in the gel at the same time. 6µl will yield the pattern and DNA concentrations shown in the picture.
Catalog No. 50-204-5809 Supplier LAMDA BIOTECH INC Supplier No. M102
100bp Quanti DNA Ladder consists of 9 fragments ranging from 100-1500 base pairs (bp). This molecular weight Marker is specially designed for identifying small DNA fragments and allowing for quantitative estimation of the DNA fragments in the gel at the same time. 6µl will yield the pattern and DNA concentrations shown in the picture.