Learn More
National Safety Apparel™ 100 Cal ArcGuard™ Nomex/Kevlar Bib Overalls
SDP
Catalog No. 17001301 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-301 Small
17-001-302 Medium
17-001-303 Large
17-001-304 X-Large
17-001-305 2X-Large
17-001-306 3X-Large
17-001-307 4X-Large
17-001-308 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-301 Supplier National Safety Apparel™ Supplier No. C45KDQESM32
Add to Cart
Add to Cart
Provides lightweight, durable, heat and flame-resistant solutions. Made of 34% aramid, 33% lyocell, 31% modacrylic, and 2% anti-static materials, Nomex™ is engineered to absorb moisture and dry quickly
  • Arc Rating: 100 cal/cm²
  • CAT 4
  • Fabric/Material: Multilayer DuPont™ Nomex™ and Kevlar™
  • FR fabric will not melt, drip, or ignite
  • FR hook and loop calf closure for easy boot entry
  • Quick disconnect and easy adjustable shoulder straps for quick entry Inherently flame resistant for FR protection
  • Side slant pockets on bib overalls
  • Expandable leg opening to fit over boots
  • Garments embroidered with arc rating Available in a convenient kit
  • Made in the USA
Specifications
Closure Type Buckle
Color Brown
Material Multi-layer DuPont™ Nomex™, Kevlar™
Seam Type Stitched
Size Small
Suspender Adjustable
Ankle Style Open
Arc Flash Rating 100 cal/cm²
Waist Style Standard
Resistance Flame
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269
Disposable Reusable