Learn More
Ted Pella Inc Grid Storage Box for 100 Grids, with a Record Card
SDP
Catalog No. NC9967927
Encompass
Intended for use in both routine grid handling and long term grid storage. Holds 100 transmission electron microscopy grids. Each of the diamond shaped compartments will accept a standard 3.05mm grid. Each position has a letter and number molded into the surface of the box. To further ease grid handling, the plastics are anti-static treated. A central access position in the lid reduces the number of grids exposed when selecting any particular grid. Dimensions: 85 x 59 x 7mm. A record card is supplied free with each SB 100 grid box. The folding card has a durable coated exterior and the interior is suitable for writing with normal pen or pencil.
Catalog No. NC9967927 Supplier Ted Pella Inc Supplier No. SB100
Intended for use in both routine grid handling and long term grid storage. Holds 100 transmission electron microscopy grids. Each of the diamond shaped compartments will accept a standard 3.05mm grid. Each position has a letter and number molded into the surface of the box. To further ease grid handling, the plastics are anti-static treated. A central access position in the lid reduces the number of grids exposed when selecting any particular grid. Dimensions: 85 x 59 x 7mm. A record card is supplied free with each SB 100 grid box. The folding card has a durable coated exterior and the interior is suitable for writing with normal pen or pencil.