Learn More
Electron Microscopy Sciences Grid Storage Box, 100 Capacity, 12/pk
SDP
Catalog No. 50930864
Encompass_Preferred
A newly designed grid storage box similar to the original LKB box. Made from a special plastic that minimizes static. Complete with grid recording card.
Catalog No. 50-930-864 Supplier Electron Microscopy Sciences Supplier No. 71156
A newly designed grid storage box similar to the original LKB box. Made from a special plastic that minimizes static. Complete with grid recording card.