Learn More
Brookfield Engineer Labs General Purpose Silicone Fluids, 100 CPS
Catalog No. NC1600904
Encompass
Brookfield Viscosity Standards provide a convenient, reliable way to verify the calibration of your Brookfield Laboratory Viscometer/Rheometer. The fluids are stored in glass bottles. Silicone viscosity standards supplied in ½ liter (1 pint) glass jars complete with a certificate of calibration.
Catalog No. NC1600904 Supplier Brookfield Engineer Labs Supplier No. 100 CPS FLUID
Brookfield Viscosity Standards provide a convenient, reliable way to verify the calibration of your Brookfield Laboratory Viscometer/Rheometer. The fluids are stored in glass bottles. Silicone viscosity standards supplied in ½ liter (1 pint) glass jars complete with a certificate of calibration.