Learn More
Chemical & Chromatography Supplies Inc 10 µm Full Flow Filter, UHMWPE Material, Effective Particulate Removal, 100 per Pack
SDP
Supplier:  Chemical & Chromatography Supplies Inc 174000
Encompass
The 10 µm Full Flow Filter is designed for high-performance liquid chromatography (HPLC) and dissolution testing. Manufactured from UHMW polyethylene, these blue filters effectively remove particulates, ensuring optimal flow rates and reducing backpressure. They are compatible with various Agilent HPLC systems and meet USP and FDA guidelines, making them ideal for diverse research applications. Supplied in packs of 100, this filter enhances component lifespan and minimizes instrument downtime.
Catalog No. NC1324999