Learn More
Analytical Sales & Services 100µL PEEK Sample Loop
Catalog No. 5000804977
Encompass
100µL PEEK Sample Loop for High Pressure Automatic Switching Valves and 9125, 9725 and 9010 Manual PEEK Injector Valves\n\nQty/Pkg: EACH
Catalog No. 50-008-04977 Supplier Analytical Sales & Services Supplier No. 490550244
100µL PEEK Sample Loop for High Pressure Automatic Switching Valves and 9125, 9725 and 9010 Manual PEEK Injector Valves\n\nQty/Pkg: EACH