Learn More
Analytical Sales & Services 100µL, PP Inserts with Standard Opening and Bottom Spring
Catalog No. 5000806591
Encompass
Advantage™ 100µL Volume, Polypropylene Inserts with Standard Opening and Bottom Spring
Catalog No. 50-008-06591 Supplier Analytical Sales & Services Supplier No. 20502CASE
Advantage™ 100µL Volume, Polypropylene Inserts with Standard Opening and Bottom Spring