Learn More
Analytical Sales & Services 100µL Slimline GC Syringe for LEAP/Gerstel CTC PAL GC Autosamplers
Catalog No. 5000806063
Encompass
100µL Slimline GC Syringe 1710 for LEAP/Gerstel CTC PAL GC Autosamplers.
26s Gauge, 51mm Length, Poinst Style AS
Qty/Pkg: EACH
Catalog No. 50-008-06063 Supplier Analytical Sales & Services Supplier No. 42030764
100µL Slimline GC Syringe 1710 for LEAP/Gerstel CTC PAL GC Autosamplers.
26s Gauge, 51mm Length, Poinst Style AS
Qty/Pkg: EACH