Learn More
Electron Microscopy Sciences Microscope Slide Boxes, Blue, 25 slides
SDP
Supplier:  Electron Microscopy Sciences 71459B
Encompass_Preferred
Molded from one piece (lid and bottom) polypropylene these slide boxes are able to accommodate either 25 or 100 standard 3x1" (75x25 mm) slides. The attached lid is held securely by a closed latch lock. Stacking feet maximize the stability when boxes are stacked up one atop the other. The boxes are unlined and the index number is printed on the bottom. Comes complete with a numbered slide index card. These boxes are available in blue, red or white.
Color: Blue
Capacity: 25 slides
Catalog No. 50-189-9601