Learn More
Electron Microscopy Sciences Microscope Slide Boxes, Red, 100 slides
SDP
Catalog No. 501899602
Encompass_Preferred
Molded from one piece (lid and bottom) polypropylene these slide boxes are able to accommodate either 25 or 100 standard 3x1" (75x25 mm) slides. The attached lid is held securely by a closed latch lock. Stacking feet maximize the stability when boxes are stacked up one atop the other. The boxes are unlined and the index number is printed on the bottom. Comes complete with a numbered slide index card. These boxes are available in blue, red or white.
Color: Red
Capacity: 100 slides
Catalog No. 50-189-9602 Supplier Electron Microscopy Sciences Supplier No. 71459R
Molded from one piece (lid and bottom) polypropylene these slide boxes are able to accommodate either 25 or 100 standard 3x1" (75x25 mm) slides. The attached lid is held securely by a closed latch lock. Stacking feet maximize the stability when boxes are stacked up one atop the other. The boxes are unlined and the index number is printed on the bottom. Comes complete with a numbered slide index card. These boxes are available in blue, red or white.
Color: Red
Capacity: 100 slides