Learn More
TA Instruments 100 Mg Class 1 Precision Weight, Weight Calibration, TGA Instruments, 100 Mg Class 1 Precision Weight
Catalog No. 5000580827 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments 100 mg Class 1 Precision Weight is used for weight calibration of TGA instruments. It ensures the accuracy of sample mass measurements for precise thermogravimetric analysis results.
Catalog No. 50-005-80827 Supplier TA Instruments Supplier No. 269931.001
Waters TA Instruments 100 mg Class 1 Precision Weight is used for weight calibration of TGA instruments. It ensures the accuracy of sample mass measurements for precise thermogravimetric analysis results.