Learn More
Leinco Technologies Inc Anti-Mouse CD1d (Clone D5) – Purified in vivo PLATINUM™ Functional Grade
SDP
Supplier:  Leinco Technologies Inc C2450100MG
Encompass_Preferred
CD1d antibody, D5, recognizes human CD1d, also known as CD1.1 and Ly-38, a 48 kDa type 1 membrane glycoprotein that belongs to the cluster of differentiation 1 (CD1) family of glycoproteins. CD1d is expressed on epithelial cells and monocyte-derived antigen presenting cells (APCs), including monocytes, DCs, and macrophages1,2. CD1d is an MHC class I-like molecule involved in the presentation of lipid antigens, such as α-galactosylceramide (α-GalCer), to a highly conserved T cell subset, named invariant natural killer T (iNKT) cells or CD1d-restricted T cells1. Following activation, iNKT cells rapidly proliferate and produce large amounts of cytokines and cytolytic mediators3,4.
Catalog No. 50-254-4247